3-Bromo-2-(2-chlorobenzyloxy)-5-methylphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (3-Bromo-2-((2-chlorobenzyl)oxy)-5-methylphenyl)boronic acid

BB-8030-1G BB-8030-250M
COBBBB-8030-1GEA 0 GBP
COBBBB-8030-1G COBBBB-8030-250M
3-Bromo-2-(2-chlorobenzyloxy)-5-methylphenylboronic acid
3-Bromo-2-(2-chlorobenzyloxy)-5-methylphenylboronic acid
Formula: C₁₄H₁₃BBrClO₃
MW: 355.42 g/mol
MDL Number: MFCD06411364
CAS Number: 849052-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR