5-Chloro-2-methyl-4-thiocyanatoaniline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2726-5G HC-2726-1G
COBBHC-2726-5GEA 0 GBP
COBBHC-2726-5G COBBHC-2726-1G
5-Chloro-2-methyl-4-thiocyanatoaniline
5-Chloro-2-methyl-4-thiocyanatoaniline
Formula: C₈H₇ClN₂S
MDL Number: MFCD12026046
CAS Number: 23530-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR