3-Chloro-2-methyl-4-thiocyanatoaniline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AN-1404-1G
AN-1404-5G
COBBAN-1404-1GEA
0
GBP
COBBAN-1404-1G
COBBAN-1404-5G
3-Chloro-2-methyl-4-thiocyanatoaniline
3-Chloro-2-methyl-4-thiocyanatoaniline
Formula:
C₈H₇ClN₂S |
MDL Number:
MFCD11855819 CAS Number: 14030-84-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...