Coniferyl alcohol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(3-Hydroxyprop-1-en-1-yl)-2-methoxyphenol

M-Clarity™ quality level = MQ100

223735-100MG 223735-1G
SIAL223735-100MGEA 136 GBP
SIAL223735-100MG SIAL223735-1G
Coniferyl alcohol, Sigma-Aldrich®
Coniferyl alcohol
Formula: C₁₀H₁₂O₃
MW: 180.20 g/mol
MDL Number: MFCD00002922
CAS Number: 458-35-5
EINECS: 207-277-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR