N,N-Dimethyl-2-(4-nitrophenoxy)ethanamine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1214-25G AN-1214-5G AN-1214-1G
COBBAN-1214-25GEA 0 GBP
COBBAN-1214-25G COBBAN-1214-5G COBBAN-1214-1G
N,N-Dimethyl-2-(4-nitrophenoxy)ethanamine
N,N-Dimethyl-2-(4-nitrophenoxy)ethanamine
Formula: C₁₀H₁₄N₂O₃
MDL Number: MFCD01410114
CAS Number: 51344-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR