N,N-Diethyl-2-(2-nitrophenoxy)ethanamine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1220-5G AN-1220-25G AN-1220-1G
COBBAN-1220-5GEA 0 GBP
COBBAN-1220-5G COBBAN-1220-25G COBBAN-1220-1G
N,N-Diethyl-2-(2-nitrophenoxy)ethanamine
N,N-Diethyl-2-(2-nitrophenoxy)ethanamine
Formula: C₁₂H₁₈N₂O₃
MDL Number: MFCD01308707
CAS Number: 96115-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR