4-(tert-Butyldimethylsilyloxy)-3,5-dichlorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-5537-1G BB-5537-5G BB-5537-250M
COBBBB-5537-1GEA 0 GBP
COBBBB-5537-1G COBBBB-5537-5G COBBBB-5537-250M
4-(tert-Butyldimethylsilyloxy)-3,5-dichlorophenylboronic acid
4-(tert-Butyldimethylsilyloxy)-3,5-dichlorophenylboronic acid
Formula: C₁₂H₁₉BCl₂O₃Si
MDL Number: MFCD12025965
CAS Number: 1150114-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR