2,6-Diformyl-3,5-dimethoxyphenol

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BD-0037-1G BD-0037-5G
COBBBD-0037-1GEA 0 GBP
COBBBD-0037-1G COBBBD-0037-5G
2,6-Diformyl-3,5-dimethoxyphenol
2,6-Diformyl-3,5-dimethoxyphenol
Formula: C₁₀H₁₀O₅
MDL Number: MFCD01632212
CAS Number: 125666-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR