5-Bromo-2-nitro-4-(trifluoromethoxy)aniline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2842-1G HC-2842-5G
COBBHC-2842-1GEA 0 GBP
COBBHC-2842-1G COBBHC-2842-5G
5-Bromo-2-nitro-4-(trifluoromethoxy)aniline
5-Bromo-2-nitro-4-(trifluoromethoxy)aniline
Formula: C₇H₄BrF₃N₂O₃
MDL Number: MFCD12756436
CAS Number: 755030-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR