N-Boc-N-isoproply 4-bromoaniline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1412-5G AN-1412-1G
COBBAN-1412-5GEA 0 GBP
COBBAN-1412-5G COBBAN-1412-1G
N-Boc-N-isoproply 4-bromoaniline
N-Boc-N-isoproply 4-bromoaniline
Formula: C₁₄H₂₀BrNO₂
MDL Number: MFCD11855823
CAS Number: 1133115-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR