5-Bromo-3-methyl-2-(2,2,2-trimethylacetamido)pyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-1174-1G PY-1174-5G
COBBPY-1174-1GEA 0 GBP
COBBPY-1174-1G COBBPY-1174-5G
5-Bromo-3-methyl-2-(2,2,2-trimethylacetamido)pyridine
5-Bromo-3-methyl-2-(2,2,2-trimethylacetamido)pyridine
Formula: C₁₁H₁₅BrN₂O
MW: 271.16 g/mol
MDL Number: MFCD09037447
CAS Number: 446299-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR