Diethoxymethylphenylsilane, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Diethoxy(methyl)(phenyl)silane

M-Clarity™ quality level = MQ100

SIAL448605-10MLEA 85.3 GBP
SIAL448605-10ML
Diethoxymethylphenylsilane, Sigma-Aldrich®
Diethoxymethylphenylsilane
Formula: C₁₁H₁₈O₂Si
MW: 210.35 g/mol
MDL Number: MFCD00048541
CAS Number: 775-56-4
EINECS: 212-275-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR