Ethyl-4-methylpyrrole-2-carboxylate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl 4-Methyl-2-pyrrolecarboxylate

AS23914G1 AS23914G25 AS23914G5
ACTIAS23914G1EA 110 GBP
ACTIAS23914G1 ACTIAS23914G25 ACTIAS23914G5
Ethyl-4-methylpyrrole-2-carboxylate
Ethyl 4-methylpyrrole-2-carboxylate
Formula: C₈H₁₁NO₂
MW: 153.1800 g/mol
MDL Number: MFCD00191668
CAS Number: 40611-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR