Ethyl pyrrole-2-carboxylate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl 1H-pyrrole-2-carboxylate

AS2075G25 AS2075G5
ACTIAS2075G25EA 0 GBP
ACTIAS2075G25 ACTIAS2075G5
Ethyl pyrrole-2-carboxylate
Ethyl pyrrole-2-carboxylate
Formula: C₇H₉NO₂
MW: 139.16 g/mol
MDL Number: MFCD00817049
CAS Number: 2199-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR