4,6-Dichloro-2-(methylthio)pyrimidine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21208G100 AS21208G25
ACTIAS21208G100EA 212 GBP
ACTIAS21208G100 ACTIAS21208G25
4,6-Dichloro-2-(methylthio)pyrimidine
4,6-Dichloro-2-(methylthio)pyrimidine
Formula: C₅H₄Cl₂N₂S
MW: 195.07 g/mol
MDL Number: MFCD00006086
CAS Number: 6299-25-8
EINECS: 228-577-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR