2-(Methylamino)-5-pyrimidineboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2-(Methylamino)pyrimidin-5-yl)boronic acid

BB-5920-1G BB-5920-250M
COBBBB-5920-1GEA 0 GBP
COBBBB-5920-1G COBBBB-5920-250M
2-(Methylamino)-5-pyrimidineboronic acid
2-(Methylamino)-5-pyrimidineboronic acid
Formula: C₅H₈BN₃O₂
MW: 152.9500 g/mol
MDL Number: MFCD16618931
CAS Number: 1033745-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR