1,3-Dimethyl-1H-pyrazol-4-carbaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1,3-Dimethyl-1H-pyrazole-4-carbaldehyde

014484-1G 014484-5G
FLUO014484-1GEA 0 GBP
FLUO014484-1G FLUO014484-5G
1,3-Dimethyl-1H-pyrazol-4-carbaldehyde
1,3-Dimethyl-1H-pyrazol-4-carbaldehyde
Formula: C₆H₈N₂O
MW: 124.1405 g/mol
MDL Number: MFCD00159635
CAS Number: 25016-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR