1-((3-Bromophenyl)sulphonyl)piperidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0151-5G OT-0151-1G
COBBOT-0151-5GEA 0 GBP
COBBOT-0151-5G COBBOT-0151-1G
1-((3-Bromophenyl)sulphonyl)piperidine
1-((3-Bromophenyl)sulphonyl)piperidine
Formula: C₁₁H₁₄BrNO₂S
MW: 304.2000 g/mol
MDL Number: MFCD07363827
CAS Number: 871269-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR