1-(4-Fluorobenzoyl)piperidine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4747-5G
CA-4747-25G
COBBCA-4747-5GEA
0
GBP
COBBCA-4747-5G
COBBCA-4747-25G
1-(4-Fluorobenzoyl)piperidine
1-(4-Fluorobenzoyl)piperidine
Formula:
C₁₂H₁₄BrNO |
MDL Number:
MFCD00174322 CAS Number: 58547-67-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...