2-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]-2-(3,4-dimethoxyphenyl)acetonitrile

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

3G-041.500 3G-041.1 3G-041.5 3G-041.10
BIOT3G-041.500EA 0 GBP
BIOT3G-041.500 BIOT3G-041.1 BIOT3G-041.5 BIOT3G-041.10
2-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]-2-(3,4-dimethoxyphenyl)acetonitrile
2-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]-2-(3,4-dimethoxyphenyl)acetonitrile
Formula: C₁₆H₁₂ClF₃N₂O₂
MDL Number: MFCD00172216
CAS Number: 338407-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR