(Triphenylstannyl)dimethylphenylsilane
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
009698-1G
009698-5G
FLUO009698-1GEA
0
GBP
FLUO009698-1G
FLUO009698-5G
(Triphenylstannyl)dimethylphenylsilane
(Triphenylstannyl)dimethylphenylsilane
Formula:
C₂₆H₂₆SiSn |
MDL Number:
MFCD03093830 CAS Number: 210362-76-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...