1-Boc-3-propylpiperazine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS12160G1 AS12160G5
ACTIAS12160G1EA 0 GBP
ACTIAS12160G1 ACTIAS12160G5
1-Boc-3-propylpiperazine
1-Boc-3-propylpiperazine
Formula: C₁₂H₂₄N₂O₂
MDL Number: MFCD08273956
CAS Number: 502649-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR