1-Boc-3-isopropylpiperazine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Boc-3-Isopropylpiperazine

AS12161G1 AS12161G5
ACTIAS12161G1EA 159 GBP
ACTIAS12161G1 ACTIAS12161G5
1-Boc-3-isopropylpiperazine
1-Boc-3-isopropylpiperazine
Formula: C₁₂H₂₄N₂O₂
MW: 228.3312 g/mol
MDL Number: MFCD08273958
CAS Number: 502649-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR