(R)-1-Boc-3-Ethylpiperazine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (R)-tert-Butyl 3-ethylpiperazine-1-carboxylate, (3R)-3-Ethylpiperazine, N1-BOC protected

AS3126G1 AS3126G5
ACTIAS3126G1EA 57 GBP
ACTIAS3126G1 ACTIAS3126G5
(R)-1-Boc-3-Ethylpiperazine
(R)-1-Boc-3-ethylpiperazine
Formula: C₁₁H₂₂N₂O₂
MW: 214.3100 g/mol
MDL Number: MFCD07772111
CAS Number: 438050-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR