4-(3-Bromophenylsulphonyl)morpholine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0152-1G OT-0152-5G
COBBOT-0152-1GEA 0 GBP
COBBOT-0152-1G COBBOT-0152-5G
4-(3-Bromophenylsulphonyl)morpholine
4-(3-Bromophenylsulphonyl)morpholine
Formula: C₁₀H₁₂BrNO₃S
MDL Number: MFCD07363828
CAS Number: 871269-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR