4-(2-Bromophenylsulphonyl)morpholine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6303-5G HC-6303-25G HC-6303-1G
COBBHC-6303-5GEA 0 GBP
COBBHC-6303-5G COBBHC-6303-25G COBBHC-6303-1G
4-(2-Bromophenylsulphonyl)morpholine
4-(2-Bromophenylsulphonyl)morpholine
Formula: C₁₀H₁₂BrNO₃S
MDL Number: MFCD09340792
CAS Number: 688798-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR