(S)-1-Boc-piperidine-3-carboxylic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (S)-1-(tert-Butoxycarbonyl)piperidine-3-carboxylic acid1-Boc-L-nipecotic acid

M-Clarity™ quality level = MQ100

SIAL681229-500MGEA 116 GBP
SIAL681229-500MG
(S)-1-Boc-piperidine-3-carboxylic acid, Sigma-Aldrich®
(S)-1-Boc-piperidine-3-carboxylic acid
Formula: C₁₁H₁₉NO₄
MW: 229.28 g/mol
MDL Number: MFCD02179172
CAS Number: 88495-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR