(S)-1-Boc-piperidine-3-carboxylic acid

Supplier: COMBI-BLOCKS INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (S)-1-(tert-Butoxycarbonyl)piperidine-3-carboxylic acid1-Boc-L-nipecotic acid

COBBOR-1733-25GEA 0 GBP
COBBOR-1733-25G
(S)-1-Boc-piperidine-3-carboxylic acid
(S)-1-Boc-piperidine-3-carboxylic acid
Formula: C₁₁H₁₉NO₄
MW: 229.28 g/mol
MDL Number: MFCD02179172
CAS Number: 88495-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR