(R)-1-Boc-piperidine-3-carboxylic acid
Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(R)-N-BOC-piperidine-3-carboxylic acid
, 1-Boc-D-nipecotic acid
, (R)-1-(tert-Butoxycarbonyl)piperidine-3-carboxylic acid
ACTIAS25131G5EA
26
GBP
ACTIAS25131G5
ACTIAS25131G25
(R)-1-Boc-piperidine-3-carboxylic acid
(R)-1-Boc-piperidine-3-carboxylic acid
Formula:
C₁₁H₁₉NO₄ MW: 229.28 g/mol |
MDL Number:
MFCD02179173 CAS Number: 163438-09-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...