(R)-(-)-3-Aminopiperidine dihydrochloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (R)-(-)-3-Piperidinamine dihydrochloride, (R)-Piperidin-3-amine dihydrochloride

AM-1746-5G AM-1746-1G
COBBAM-1746-5GEA 0 GBP
COBBAM-1746-5G COBBAM-1746-1G
(R)-(-)-3-Aminopiperidine dihydrochloride
(R)-(-)-3-Aminopiperidine dihydrochloride
Formula: C₅H₁₄Cl₂N₂
MW: 173.08 g/mol
MDL Number: MFCD06799458
CAS Number: 334618-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR