1-(3-Bromophenylsulphonyl)pyrrolidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-((3-Bromophenyl)sulphonyl)pyrrolidine

OT-0153-5G OT-0153-1G
COBBOT-0153-5GEA 0 GBP
COBBOT-0153-5G COBBOT-0153-1G
1-(3-Bromophenylsulphonyl)pyrrolidine
1-(3-Bromophenylsulphonyl)pyrrolidine
Formula: C₁₀H₁₂BrNO₂S
MW: 290.1768 g/mol
MDL Number: MFCD07363829
CAS Number: 214210-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR