2-Bromo-6-(3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl)pyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-8035-1G PY-8035-5G
COBBPY-8035-1GEA 0 GBP
COBBPY-8035-1G COBBPY-8035-5G
2-Bromo-6-(3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl)pyridine
2-Bromo-6-(3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl)pyridine
Formula: C₁₀H₇BrF₃N₃
MDL Number: MFCD12546619
CAS Number: 1187386-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR