3-Methyl-2-(tributylstannyl)thiophene

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS51231G1 AS51231G5
ACTIAS51231G1EA 282 GBP
ACTIAS51231G1 ACTIAS51231G5
3-Methyl-2-(tributylstannyl)thiophene
3-Methyl-2-(tributylstannyl)thiophene
Formula: C₁₇H₃₂SSn
MDL Number: MFCD09025807
CAS Number: 166766-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR