Victoria blue B, dark brown powder C.I. 44045
Supplier: MP Biomedicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
N-(4-((4-(Dimethylamino)phenyl)(4-(phenylamino)naphthalen-1-yl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanaminium chloride
ICNA0215829125EA
43.3
GBP
ICNA0215829125
Victoria blue B, dark brown powder C.I. 44045
Victoria Blue B
Victoria Blue B is used as a nuclear stain in cytology/histochemical applications. Victoria Blue B may be used in the quantitative determination of phospholipids.
More soluble in the lower aliphatic alcohols; slightly soluble in water.
Caution: Research Use Only (RUO).
Formula:
C₃₃H₃₂ClN₃ MW: 506.803 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011878 CAS Number: 2580-56-5 EINECS: 219-943-6 |
Specification Test Results
Identity Test | Passes |
UV/Vis: λmax (ethanol) | Report Resut |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...