Victoria pure blue bo

Supplier: EHRENSTORFER GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Basic Blue 7N-(4-((4-(Diethylamino)phenyl)(4-(ethylamino)naphthalen-1-yl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium chloride

EHERC10424000EA 357 GBP
EHERC10424000
Victoria pure blue bo
Victoria Pure Blue Bo
Formula: C₃₃H₄₀ClN₃
MW: 514.1438 g/mol
MDL Number: MFCD00011877
CAS Number: 2390-60-5
EINECS: 219-232-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR