2-Adamantyl-1-benzyloxy-4-bromobenzene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-[2-(Benzyloxy)-5-bromophenyl]adamantane

OT-0466-1G OT-0466-5G
COBBOT-0466-1GEA 0 GBP
COBBOT-0466-1G COBBOT-0466-5G
2-Adamantyl-1-benzyloxy-4-bromobenzene
2-Adamantyl-1-benzyloxy-4-bromobenzene
Formula: C₂₃H₂₅BrO
MDL Number: MFCD09258646
CAS Number: 736992-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR