4-(4-Boc-1-piperazinylcarbonyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (4-(4-(tert-Butoxycarbonyl)piperazine-1-carbonyl)phenyl)boronic acid, 1-(1,1-Dimethylethyl) 4-(4-boronobenzoyl)-1-piperazinecarboxylate

BB-8909-5G BB-8909-1G
COBBBB-8909-5GEA 0 GBP
COBBBB-8909-5G COBBBB-8909-1G
4-(4-Boc-1-piperazinylcarbonyl)phenylboronic acid
4-(4-Boc-1-piperazinylcarbonyl)phenylboronic acid
Formula: C₁₆H₂₃BN₂O₅
MW: 334.1752 g/mol
MDL Number: MFCD12025996
CAS Number: 1150114-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR