Victoria blue B 0.1% in 2-propanol

Supplier: DISTRIBUTOR UK
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Basic blue 26N-(4-((4-(Dimethylamino)phenyl)(4-(phenylamino)naphthalen-1-yl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanaminium chloride

DIUKFL2220EA 32.2 GBP
DIUKFL2220
Victoria blue B 0.1% in 2-propanol
Victoria Blue B
Formula: C₃₃H₃₂ClN₃
MW: 506.803 g/mol
MDL Number: MFCD00011878
CAS Number: 2580-56-5
EINECS: 219-943-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR