2-(1H-Pyrazol-1-yl)benzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20629G1 AS20629G25 AS20629G5
ACTIAS20629G1EA 37 GBP
ACTIAS20629G1 ACTIAS20629G25 ACTIAS20629G5
2-(1H-Pyrazol-1-yl)benzoic acid
2-(1H-Pyrazol-1-yl)benzoic acid
Formula: C₁₀H₈N₂O₂
MDL Number: MFCD03086184
CAS Number: 55317-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR