5-Bromo-2-(piperidin-1-yl)pyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PY-7943-5G
PY-7943-1G
PY-7943-25G
COBBPY-7943-5GEA
0
GBP
COBBPY-7943-5G
COBBPY-7943-1G
COBBPY-7943-25G
5-Bromo-2-(piperidin-1-yl)pyridine
5-Bromo-2-(piperidin-1-yl)pyridine
Formula:
C₁₀H₁₃BrN₂ |
MDL Number:
MFCD00234654 CAS Number: 24255-95-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...