1-(5-Bromo-4-methyl-2-pyridinyl)piperazine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-8055-5G OT-8055-25G OT-8055-1G
COBBOT-8055-5GEA 0 GBP
COBBOT-8055-5G COBBOT-8055-25G COBBOT-8055-1G
1-(5-Bromo-4-methyl-2-pyridinyl)piperazine
1-(5-Bromo-4-methyl-2-pyridinyl)piperazine
Formula: C₁₀H₁₄BrN₃
MDL Number: MFCD09260438
CAS Number: 944582-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR