6-Bromo-N-methylpicolinamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PY-7221-1G
PY-7221-5G
COBBPY-7221-1GEA
0
GBP
COBBPY-7221-1G
COBBPY-7221-5G
6-Bromo-N-methylpicolinamide
6-Bromo-N-methylpicolinamide
Formula:
C₇H₇BrN₂O |
MDL Number:
MFCD10699703 CAS Number: 337535-94-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...