6-Bromopicolinamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Bromopyridine-2-carboxamide

CA-4203-5G CA-4203-25G CA-4203-1G
COBBCA-4203-5GEA 0 GBP
COBBCA-4203-5G COBBCA-4203-25G COBBCA-4203-1G
6-Bromopicolinamide
6-Bromopicolinamide
Formula: C₆H₅BrN₂O
MW: 201.02 g/mol
MDL Number: MFCD00234154
CAS Number: 25194-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR