5-Bromopicolinamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Bromopyridine-2-carboxamide

CA-4207-5G CA-4207-1G
COBBCA-4207-5GEA 0 GBP
COBBCA-4207-5G COBBCA-4207-1G
5-Bromopicolinamide
5-Bromopicolinamide
Formula: C₆H₅BrN₂O
MW: 201.0200 g/mol
MDL Number: MFCD04066715
CAS Number: 90145-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR