Tetrahydropyran-2-carboxylic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Tetrahydro-2H-pyran-2-carboxylic acid
AS20728G1
AS20728G5
ACTIAS20728G1EA
0
GBP
ACTIAS20728G1
ACTIAS20728G5
Tetrahydropyran-2-carboxylic acid
Tetrahydropyran-2-carboxylic acid
Formula:
C₆H₁₀O₃ MW: 130.1418 g/mol |
MDL Number:
MFCD07779239 CAS Number: 51673-83-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...