8-Fluoro-4-quinolinol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 8-Fluoroquinolin-4-ol, 8-Fluoro-4-hydroxyquinoline

AS20939G1 AS20939G25 AS20939G5
ACTIAS20939G1EA 23 GBP
ACTIAS20939G1 ACTIAS20939G25 ACTIAS20939G5
8-Fluoro-4-quinolinol
8-Fluoro-4-quinolinol
Formula: C₉H₆FNO
MW: 163.15 g/mol
MDL Number: MFCD00272395
CAS Number: 63010-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR