3-(2'-Chlorobenzyloxy)-2,6-difluorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-8100-1G BB-8100-250M
COBBBB-8100-1GEA 0 GBP
COBBBB-8100-1G COBBBB-8100-250M
3-(2'-Chlorobenzyloxy)-2,6-difluorophenylboronic acid
3-(2'-Chlorobenzyloxy)-2,6-difluorophenylboronic acid
Formula: C₁₃H₁₀BClF₂O₃
MDL Number: MFCD08276771
CAS Number: 870778-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR