5-Chloro-2-methyl-3-nitrophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-3825-1G BB-3825-5G
COBBBB-3825-1GEA 0 GBP
COBBBB-3825-1G COBBBB-3825-5G
5-Chloro-2-methyl-3-nitrophenylboronic acid
5-Chloro-2-methyl-3-nitrophenylboronic acid
Formula: C₇H₇BClNO₄
MDL Number: MFCD09027246
CAS Number: 957060-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR