4-(4-Fluoro-2-nitrophenyl)morpholine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
017159-1G
017159-5G
FLUO017159-1GEA
0
GBP
FLUO017159-1G
FLUO017159-5G
4-(4-Fluoro-2-nitrophenyl)morpholine
4-(4-Fluoro-2-nitrophenyl)morpholine
Formula:
C₁₀H₁₁FN₂O₃ MW: 226.20 g/mol |
MDL Number:
MFCD00798369 CAS Number: 238418-75-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...