4-(4-Fluoro-2-nitrophenyl)morpholine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1202-5G AN-1202-25G AN-1202-1G
COBBAN-1202-5GEA 0 GBP
COBBAN-1202-5G COBBAN-1202-25G COBBAN-1202-1G
4-(4-Fluoro-2-nitrophenyl)morpholine
4-(4-Fluoro-2-nitrophenyl)morpholine
Formula: C₁₀H₁₁FN₂O₃
MW: 226.20 g/mol
MDL Number: MFCD00798369
CAS Number: 238418-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR